Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261718.1 | NP_001248647.1 | 452 | 88 | ||
XM_015128385.2 | XP_014983871.1 | 531 | 88 | |||
XM_015128386.2 | XP_014983872.1 | 820 | 88 | |||
XM_015128387.2 | XP_014983873.1 | 531 | 88 | |||
GenBank mRNA | JU322282.1 | - | 503 | 88 | ||
JV047122.1 | - | 452 | 88 | |||
JV652072.1 | - | 531 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |