Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261789.1 | NP_001248718.1 | 168 | 115 | ||
GenBank mRNA | JU336914.1 | - | 163 | 115 | ||
JU477650.1 | - | 86 | 115 | |||
JV048581.1 | - | 168 | 115 | |||
JV664470.1 | - | 171 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |