Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265820.1 | NP_001252749.1 | 82 | 85 | ||
GenBank mRNA | JU336486.1 | - | 120 | 85 | ||
JU477298.1 | - | 110 | 85 | |||
JV667482.1 | - | 82 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |