Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092811.4 | XP_001092811.2 | 883 | 106 | ||
XM_015137942.2 | XP_014993428.2 | 915 | 106 | |||
GenBank mRNA | JU336257.1 | - | 863 | 106 | ||
JU477090.1 | - | 810 | 106 | |||
JV048167.1 | - | 867 | 106 | |||
JV048168.1 | - | 863 | 106 | |||
JV703397.1 | - | 888 | 106 | |||
JV703398.1 | - | 884 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |