Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261593.1 | NP_001248522.1 | 335 | 61 | ||
XM_015141543.2 | XP_014997029.2 | 913 | 61 | |||
XM_015141544.2 | XP_014997030.2 | 1030 | 61 | |||
XM_015141545.2 | XP_014997031.2 | 776 | 61 | |||
XM_015141546.2 | XP_014997032.2 | 1051 | 61 | |||
XM_015141547.2 | XP_014997033.2 | 422 | 61 | |||
XM_015141549.2 | XP_014997035.2 | 474 | 61 | |||
XM_028849162.1 | XP_028704995.1 | 740 | 61 | |||
XM_028849164.1 | XP_028704997.1 | 839 | 61 | |||
XM_028849165.1 | XP_028704998.1 | 1022 | 61 | |||
GenBank mRNA | JU321103.1 | - | 391 | 61 | ||
JU470894.1 | - | 335 | 61 | |||
JU476825.1 | - | 327 | 61 | |||
JU476826.1 | - | 455 | 61 | |||
JV043445.1 | - | 335 | 61 | |||
JV565019.1 | - | 406 | 61 | |||
JV625975.1 | - | 454 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |