Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261336.1 | NP_001248265.1 | 487 | 67 | ||
XM_015131358.2 | XP_014986844.1 | 785 | 67 | |||
XM_015131360.2 | XP_014986846.1 | 434 | 67 | |||
GenBank mRNA | AY072879.1 | - | 204 | 67 | ||
JU335913.1 | - | 528 | 67 | |||
JU476790.1 | - | 487 | 67 | |||
JV658753.1 | - | 578 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |