Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266219.2 | NP_001253148.1 | 297 | 80 | ||
GenBank mRNA | JU320282.1 | - | 309 | 80 | ||
JU470709.1 | - | 304 | 80 | |||
JV044801.1 | - | 300 | 80 | |||
JV675335.1 | - | 286 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |