Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193769.1 | NP_001180698.1 | 273 | 114 | ||
GenBank mRNA | JU320593.1 | - | 253 | 114 | ||
JU472064.1 | - | 224 | 114 | |||
JV045859.1 | - | 258 | 114 | |||
JV528696.1 | - | 237 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |