Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113321.4 | XP_001113321.1 | 2393 | 62 | ||
XM_001113411.4 | XP_001113411.1 | 2529 | 62 | |||
XM_028835033.1 | XP_028690866.1 | 2529 | 62 | |||
XM_028835034.1 | XP_028690867.1 | 407 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |