Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001110411.4 | XP_001110411.1 | 1400 | 74 | ||
GenBank mRNA | JU325736.1 | - | 1354 | 74 | ||
JU491323.1 | - | 1357 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |