Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266267.1 | NP_001253196.1 | 376 | 84 | ||
GenBank mRNA | JU320144.1 | - | 369 | 84 | ||
JU472559.1 | - | 376 | 84 |