Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266972.1 | NP_001253901.1 | 583 | 103 | ||
XM_015127226.2 | XP_014982712.1 | 955 | 103 | |||
GenBank mRNA | JU320806.1 | - | 577 | 103 | ||
JV044452.1 | - | 583 | 103 | |||
JV669127.1 | - | 409 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |