Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RORA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100971.4 | XP_001100971.2 | 333 | 85 | ||
XM_015142369.2 | XP_014997855.2 | 661 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta Inventoried |