Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145282.2 | XP_015000768.2 | 1065 | 84 | ||
XM_015145283.2 | XP_015000769.2 | 1104 | 84 | |||
XM_028852634.1 | XP_028708467.1 | 952 | 84 | |||
GenBank mRNA | JV564995.1 | - | 939 | 84 | ||
JV704520.1 | - | 947 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |