Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091918.4 | XP_001091918.3 | 628 | 58 | ||
XM_015111418.2 | XP_014966904.2 | 561 | 58 | |||
XM_028831266.1 | XP_028687099.1 | 951 | 58 | |||
XM_028831267.1 | XP_028687100.1 | 892 | 58 | |||
GenBank mRNA | JV043961.1 | - | 567 | 58 | ||
JV635425.1 | - | 567 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |