Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260711.1 | NP_001247640.1 | 1083 | 89 | ||
GenBank mRNA | JU336759.1 | - | 1453 | 89 | ||
JU477523.1 | - | 1083 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |