Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEPSIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836916.1 | XP_028692749.1 | 430 | 72 | ||
XM_028836917.1 | XP_028692750.1 | 411 | 72 | |||
XM_028836918.1 | XP_028692751.1 | 411 | 72 | |||
XM_028836919.1 | XP_028692752.1 | 333 | 72 | |||
XM_028836921.1 | XP_028692754.1 | 430 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |