Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266207.1 | NP_001253136.1 | 366 | 63 | ||
XM_015114522.2 | XP_014970008.1 | 790 | 63 | |||
XM_015114524.2 | XP_014970010.1 | 500 | 63 | |||
XM_015114525.2 | XP_014970011.1 | 1130 | 63 | |||
GenBank mRNA | JU319717.1 | - | 133 | 63 | ||
JV565117.1 | - | 366 | 63 | |||
JV703597.1 | - | 382 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta Inventoried |