Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261065.1 | NP_001247994.1 | 550 | 88 | ||
GenBank mRNA | JU334515.1 | - | 550 | 88 | ||
JV684155.1 | - | 577 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |