Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261590.1 | NP_001248519.1 | 552 | 72 | ||
XM_015149745.2 | XP_015005231.1 | 462 | 72 | |||
GenBank mRNA | JU336514.1 | - | 444 | 72 | ||
JU477325.1 | - | 571 | 72 | |||
JV048319.1 | - | 552 | 72 | |||
JV662341.1 | - | 571 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |