Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266524.1 | NP_001253453.1 | 503 | 65 | ||
GenBank mRNA | JU320820.1 | - | 465 | 65 | ||
JU473750.1 | - | 438 | 65 | |||
JV047421.1 | - | 503 | 65 | |||
JV471922.1 | - | 358 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |