Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089176.4 | XP_001089176.1 | 415 | 145 | ||
XM_028841455.1 | XP_028697288.1 | 463 | 145 | |||
GenBank mRNA | JV062849.1 | - | 283 | 145 | ||
JV653246.1 | - | 213 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |