Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132280.2 | XP_014987766.1 | 687 | 94 | ||
GenBank mRNA | JU322237.1 | - | 675 | 94 | ||
JU471322.1 | - | 409 | 94 | |||
JV635343.1 | - | 696 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |