Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257898.1 | NP_001244827.1 | 125 | 61 | ||
XM_028846852.1 | XP_028702685.1 | 61 | ||||
GenBank mRNA | JU320735.1 | - | 125 | 61 | ||
JV629832.1 | - | 63 | 61 | |||
JV662449.1 | - | 133 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |