Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266489.1 | NP_001253418.1 | 358 | 80 | ||
GenBank mRNA | JU322430.1 | - | 358 | 80 | ||
JV704594.1 | - | 381 | 80 | |||
M96553.1 | - | 323 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |