Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257794.1 | NP_001244723.1 | 318 | 73 | ||
GenBank mRNA | JU334765.1 | - | 318 | 73 | ||
JU475806.1 | - | 357 | 73 | |||
JV652911.1 | - | 364 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |