Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142361.2 | XP_014997847.1 | 2528 | 122 | ||
XM_015142362.2 | XP_014997848.1 | 2572 | 122 | |||
XM_015142364.2 | XP_014997850.1 | 2408 | 122 | |||
XM_015142365.2 | XP_014997851.1 | 2384 | 122 | |||
XM_015142366.2 | XP_014997852.1 | 2412 | 122 | |||
XM_015142368.2 | XP_014997854.1 | 2528 | 122 | |||
GenBank mRNA | JU322771.1 | - | 2334 | 122 | ||
JU477218.1 | - | 2512 | 122 | |||
JV723423.1 | - | 2324 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |