Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261320.1 | NP_001248249.1 | 3539 | 63 | ||
XM_015149484.2 | XP_015004970.1 | 3565 | 63 | |||
XM_015149493.2 | XP_015004979.1 | 3713 | 63 | |||
XM_015149498.2 | XP_015004984.1 | 3491 | 63 | |||
XM_028835532.1 | XP_028691365.1 | 3394 | 63 | |||
GenBank mRNA | JU321966.1 | - | 3545 | 63 | ||
JU476687.1 | - | 3602 | 63 | |||
JV047863.1 | - | 3539 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |