Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261320.1 | NP_001248249.1 | 3801 | 73 | ||
XM_015149484.2 | XP_015004970.1 | 3827 | 73 | |||
XM_015149498.2 | XP_015004984.1 | 3753 | 73 | |||
XM_028835532.1 | XP_028691365.1 | 3656 | 73 | |||
GenBank mRNA | JU321966.1 | - | 3807 | 73 | ||
JU476687.1 | - | 3864 | 73 | |||
JV047863.1 | - | 3801 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |