Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266625.1 | NP_001253554.1 | 545 | 131 | ||
XM_015115449.2 | XP_014970935.1 | 572 | 131 | |||
GenBank mRNA | JU328134.1 | - | 433 | 131 | ||
JU475916.1 | - | 451 | 131 | |||
JV043389.1 | - | 545 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |