Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NETO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266059.1 | NP_001252988.1 | 663 | 91 | ||
XM_015126052.2 | XP_014981538.1 | 1056 | 91 | |||
XM_015126053.2 | XP_014981539.1 | 1035 | 91 | |||
XM_015126054.2 | XP_014981540.1 | 1035 | 91 | |||
XM_015126055.2 | XP_014981541.1 | 933 | 91 | |||
XM_015126056.2 | XP_014981542.1 | 1056 | 91 | |||
XM_015126058.2 | XP_014981544.1 | 1035 | 91 | |||
GenBank mRNA | JU320170.1 | - | 693 | 91 | ||
JU336561.1 | - | 672 | 91 | |||
JU477374.1 | - | 663 | 91 | |||
JU477375.1 | - | 642 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |