Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096429.4 | XP_001096429.1 | 3797 | 99 | ||
GenBank mRNA | JU470601.1 | - | 3706 | 99 | ||
JV713565.1 | - | 826 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |