Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257778.1 | NP_001244707.1 | 400 | 70 | ||
XM_015119780.2 | XP_014975266.1 | 435 | 70 | |||
XM_028835738.1 | XP_028691571.1 | 770 | 70 | |||
XM_028835740.1 | XP_028691573.1 | 435 | 70 | |||
GenBank mRNA | JU332253.1 | - | 400 | 70 | ||
JU472931.1 | - | 403 | 70 | |||
JV043835.1 | - | 399 | 70 | |||
JV529778.1 | - | 384 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |