Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104870.4 | XP_001104870.1 | 4178 | 105 | ||
XM_015126202.2 | XP_014981688.1 | 2029 | 105 | |||
GenBank mRNA | JU326539.1 | - | 639 | 105 | ||
JU326540.1 | - | 639 | 105 | |||
JV636233.1 | - | 1015 | 105 | |||
JV717360.1 | - | 651 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |