Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU326548.1 | - | 1156 | 68 | ||
JU326549.1 | - | 1156 | 68 | |||
JV723274.1 | - | 1129 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |