Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106130.4 | XP_001106130.2 | 1013 | 76 | ||
XM_015135357.2 | XP_014990843.2 | 1022 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |