Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194269.2 | NP_001181198.1 | 611 | 69 | ||
GenBank mRNA | JU321236.1 | - | 575 | 69 | ||
JU473643.1 | - | 561 | 69 | |||
JV045771.1 | - | 538 | 69 | |||
JV637107.1 | - | 579 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |