Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100171.4 | XP_001100171.3 | 197 | 102 | ||
XM_002806209.3 | XP_002806255.2 | 437 | 102 | |||
XM_028842510.1 | XP_028698343.1 | 334 | 102 | |||
GenBank mRNA | JU337313.1 | - | 209 | 102 | ||
JU478015.1 | - | 139 | 102 | |||
JV048826.1 | - | 292 | 102 | |||
JV666670.1 | - | 295 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |