Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266608.1 | NP_001253537.1 | 475 | 65 | ||
XM_015120237.2 | XP_014975723.1 | 367 | 65 | |||
XM_028835489.1 | XP_028691322.1 | 1525 | 65 | |||
XM_028835491.1 | XP_028691324.1 | 1414 | 65 | |||
GenBank mRNA | JU475398.1 | - | 475 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |