Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147516.2 | XP_015003002.1 | 595 | 74 | ||
GenBank mRNA | JU326950.1 | - | 257 | 74 | ||
JV710793.1 | - | 278 | 74 |