Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194705.3 | NP_001181634.2 | 540 | 55 | ||
XM_015124422.2 | XP_014979908.2 | 640 | 55 | |||
GenBank mRNA | JV664964.1 | - | 186 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |