Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145492.2 | XP_015000978.1 | 702 | 142 | ||
XM_028852711.1 | XP_028708544.1 | 702 | 142 | |||
XM_028852712.1 | XP_028708545.1 | 611 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |