Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257793.1 | NP_001244722.1 | 836 | 110 | ||
GenBank mRNA | JU320000.1 | - | 836 | 110 | ||
JU471632.1 | - | 1123 | 110 | |||
JV043422.1 | - | 1049 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |