Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144993.2 | XP_015000479.2 | 3468 | 79 | ||
GenBank mRNA | JV582772.1 | - | 1103 | 79 | ||
JV617637.1 | - | 1114 | 79 |