Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260725.1 | NP_001247654.1 | 1294 | 66 | ||
XM_015118357.2 | XP_014973843.1 | 1457 | 66 | |||
XM_015118365.2 | XP_014973851.1 | 1816 | 66 | |||
XM_028830473.1 | XP_028686306.1 | 1379 | 66 | |||
XM_028830503.1 | XP_028686336.1 | 1578 | 66 | |||
XM_028830601.1 | XP_028686434.1 | 1625 | 66 | |||
GenBank mRNA | JU322406.1 | - | 1294 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |