Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143828.2 | XP_014999314.2 | 1118 | 111 | ||
XM_028851594.1 | XP_028707427.1 | 945 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |