Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843828.1 | XP_028699661.1 | 1638 | 124 | ||
GenBank mRNA | JV716907.1 | - | 573 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |