Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266340.1 | NP_001253269.1 | 396 | 107 | ||
GenBank mRNA | JU323187.1 | - | 366 | 107 | ||
JU475075.1 | - | 396 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |