Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265942.2 | NP_001252871.1 | 341 | 90 | ||
GenBank mRNA | JU336359.1 | - | 330 | 90 | ||
JU477179.1 | - | 340 | 90 | |||
JV048235.1 | - | 341 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |