Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194341.2 | NP_001181270.1 | 242 | 58 | ||
GenBank mRNA | JV704188.1 | - | 221 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |